Learn More
Contec™ SATWIPES™ Presaturated Wipes

Prepare for a variety of cleanrooms. The SATWIPES™Presaturated Wipes are offered in various options to help tackle general-purpose cleaning in ISO Class 5 to ISO Class 8 environments.
Brand: Contec™ SAT-C3-AC
30.00 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
- Durable, presaturated wipes with a choice of solvent saturations on perforated rolls
- Polyester Nonwoven are ideal for critical applications in ISO Class 5 environments
- SATWIPES with Polyester/Cellulose Nonwoven Wipes offer strength and low extractables for use in less critical applications in ISO Class 6 to 8 environments
- Designed for general-purpose cleaning and surface preparation and for wiping process tools and other equipment and for use prior to cleanroom entry
Order Info
Packaged one hundred 6 x 9 in. (15 x 23cm) wipers on a perforated roll. Most SATWipes cases contain 12 presaturated rolls plus one canister; 19-150-0711 and -0712 packed 18 refills per case without a canister. Custom-blend substrates and additional wiper sizes available through your Fisher Safety Customer Service Representative. Empty canisters for wipes is available: 19-171-955.

Specifications
Pre-saturated Wipe | |
15.24 cm | |
22.86 cm | |
Not Autoclavable | |
Single-use | |
Excellent wipes for less critical applications, compatible with ISO class 6 environments and wipes down prior to entry into cleanroom | |
1-ply | |
Non-sterile |
6 in. | |
9 in. | |
Polyester | |
ISO Class 5 and 6 | |
15 x 23 cm (6 x 9 in.) | |
1200/Cs. | |
100% Acetone |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P370+P378-In case of fire: Use appropriate media to extinguish.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : Acetone
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1055806-0-0-1 | Certificate of Quality | 16/05/2024 | SAT-C3-AC |
1053995-0-0-1 | Certificate of Quality | 29/03/2024 | SAT-C3-AC |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.