Learn More
TFA Dostmann Digital Thermo-Hygrometer
This modern ultra-flat glossy digital thermo-hygrometer is an ideal instrument for monitoring the indoor climate. You can easily keep an eye on temperature and humidity. If a smiley icon appears, the values are ideal. The highest and lowest values are stored and displayed at the push of a button.
Brand:  TFA Dostmann 30.5027.02
Product Code. 16436840
  • 11.50 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
At least two thirds of a person's lifetime are spent indoors. Whether we feel well and comfortably at home depends to a great extent on the quality of the indoor air. Rooms that are too humid favour mould growth, but overly dry air can be harmful to our health and pets, plants, wooden floors and antique furniture will also suffer. This modern ultra-flat glossy thermo-hygrometer is an ideal instrument for monitoring the indoor climate. You can easily keep an eye on temperature and humidity and regulate it by carefully heating and ventilating when the values are outside the comfort-level zone. This creates a pleasant and healthy living environment and even saves on heating costs. If a smiley icon appears, the values are ideal. The highest and lowest values are stored and displayed at the push of a button.
  • For a healthy indoor climate
  • Monitoring of indoor temperature and humidity
  • Max.-min. values
  • With Smiley as a comfort level indicator
  • Ultra-flat, high-gloss design
Specifications
Specifications
Digital Thermohygrometer
20 to 95%
0°C to 50°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
TFA Dostmann Digital Thermo-Hygrometer > Temperature Range: 0°C to 50°C