Learn More
Corning™ 384-Well Low-Volume Solid Microplates
Same low reagent volumes as 1536-well plates without the need for new instrumentation
Brand:  Corning™ 3674
 View more versions of this product
Product Code. 10699921
Description
Description
Polystyrene plates feature a unique square-to-round well design with a conical-shaped bottom that provides easier access and imaging for superior assay performance.
  • Bottom reduces fluid and air entrapment, greater signal-to-noise ratio and reduced FP signal (mP) when working with very low volumes
  • Each well has a raised rim to reduce cross-contamination during assay and liquid-handling process
  • Choose black plates for fluorescence applications or white for luminescence applications
High-Binding Surface Plates
  • Ideal for immunoassays
  • Can bind medium (>10kDa) and large biomolecules that possess ionic groups and/or hydrophobic regions
  • Working volume: 2 to 20μL
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No Barcode
Without Lid
White
50 μL
5 to 40 μL
Non-treated
Provides easier access and imaging for superior assay performance, fluorescence applications, reduce cross-contamination during assay and liquid-handling process
White
Standard
16 x 24
Polystyrene
384
Non-sterile
Round
100/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ 384-Well Low-Volume Solid Microplates > Round bottom; White; Nontreated surface