Learn More
Corning™ Nonsterile Polyester Microplate Sealing Tape
GREENER_CHOICE
Provides well-to-well sealing across entire microplate to eliminate edge effect in sensitive ELISA assays. Corning™ Nonsterile Polyester Microplate Sealing Tape is suitable for sealing tissue culture mmicroplates, for short term storage incubation, for transport and the containment of solutions.
Brand:  Corning™ 4612
380.95 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Product Code. 15379393
  • 417.04 € / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Easy application and removal for short- and long-term storage
  • Provides tight seal to minimize evaporation and condensation
  • Aluminum sealing tape (Cat. Nos. 6569 and 6570) is suitable for use between -80°C and 150°C, is not transparent, and is pierceable.
  • Breathable sealing tape (Cat. No. 3345) allows gas exchange across the surface.
  • Universal Optical sealing tape (Cat. No. 6575) is suitable for use between -70°C and 100°C, and is transparent.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transparent
Non-sterile
100/Pk.
-40°C to +120°C
Polyester
500/Cs.
Microplate Sealing Tape
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Nonsterile Polyester Microplate Sealing Tape > Polyester