Learn More
Thermo Scientific™ Sealing Tape for 96-Well Plates
Clear, self-sticking sealing tape for standard microplates.
Brand: Thermo Scientific™ 15036
118.58 EUR valid until 2025-12-31
Use promo code "24288" to get your promotional price.
Description
Clear, self-sticking sealing tape for standard microplates.
Thermo Scientific Microplate Sealing Tape is a package of pre-cut, pressure-sensitive sheets for sealing the surface of standard 96-well or 384-well microplates during incubation steps.Features of Microplate Sealing Tape:
• Pressure-sensitive—position the tape over plate and press to seal; easy to remove
• Separate sheets—each sheet of tape is supplied on its own easy-to-peal backing (liner)
• Durable and inert—film thickness provides support and has a thin layer of acrylic adhesive
Use the disposable sheets to seal and reseal a microplate two or three times for incubation steps in an ELISA or protein assay procedure. Seal plates quickly and easily to allow mixing or prevent evaporation during assay incubation steps.
Specifications
Clear | |
Non-sterile | |
100 sheets | |
Pressure Sensitive |
Polyester | |
96/384 | |
Film, Seal, or Applicator | |
96/384-well Plate |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.