Learn More
Corning™ Universal Fit Pipet Tips: Stack Rack, Nonsterile
GREENER_CHOICE
Designed to assure a reliable fit with major brand pipetters
Brand:  Corning™ 4803
315.30 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
 View more versions of this product
Product Code. 10307751
  • 331.89 € / Pack of 4800
Estimated Shipment: 04-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Corning universal fit tips are designed to provide a reliable fit with most major brand pipettors. (A Pipet Tip Compatibility Guide can be requested or downloaded from the Corning website).
  • Beveled orifice helps ensure accurate fluid delivery.
  • 1-200 μL universal fit tips are graduated at the 250 μL, 500 μL, and 1,000 μL volumes.
  • Select from three packaging options: Stack rack tips feature a stack of five racks, each containing 96 tips, for a total of 480 tips in a space-saving design. Bulk packed tips are nonsterile and economical.
  • RNase-/DNase-free.
  • Nonpyrogenic.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 to 200 μL
Natural
Round
400 Tips x 10 Racks/Cs.
Polypropylene
Non-sterile
Rack
400 Tips/Stack Rack
Yes
10, 50, 100μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Universal Fit Pipet Tips: Stack Rack, Nonsterile > Natural; 400 Tips/Rack; 10 Racks/Cs; No; 1-200 μL