missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Gel Loading Buffer II (Denaturing PAGE)

A 1–2X solution of 95% Formamide, 18 mM EDTA, and 0.025% SDS, Xylene Cyanol, and Bromophenol Blue.

Brand:  Invitrogen™ AM8547

 View more versions of this product

Product Code. 10341475

  • 227.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

A 1–2X solution of 95% Formamide, 18 mM EDTA, and 0.025% SDS, Xylene Cyanol, and Bromophenol Blue. Supplied in one 1.4 mL bottle. All Invitrogen™ Gel Loading Solutions are rigorously tested for non-specific endonuclease activity, exonuclease activity, RNase activity, and for functionality. These are the same solutions we use in-house and in our kits.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at –20°C.
Bottle
Liquid
Ambion™
Room Temperature
Sample Loading Buffers
Bromophenol Blue
Denaturing Polyacrylamide Gels
10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2771042 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM8547
Lot Number2821239 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberAM8547
Lot Number2888184 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberAM8547
Lot Number2919201 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberAM8547
Lot Number01339658 Certificate TypeCertificate of Analysis Date30/06/2023 Catalog NumberAM8547
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Gel Loading Buffer II (Denaturing PAGE) > 10mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.