Learn More
Corning™ Thermowell™ 96-Well Polypropylene PCR Microplates
Constructed of medical grade polypropylene to accommodate wide temperature changes that might occur from storage to thermal cycling. Corning™ Thermowell™ 96-Well Polypropylene PCR Microplates are ideal for the polyerase chain reaction (PCR) method of DNA amplification.
Brand:  Corning™ 3758
 View more versions of this product
Product Code. 10363173
  • 608.83 € / Pack of 50
Estimated Shipment: 05-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning Thermowell GOLD 96-well PCR microplates are offered in four formats to ensure maximum flexibility and a perfect match for your applications. The original Thermowell 96-well PCR microplates are universal fit and can be cut into 3 x 8 well segments.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Includes Barcode
Standard
8 x 12
Polypropylene
Non-sterile
Conical Bottom (V-well)
2 generic bar code labels per plate
10/Pk.
DNase and RNase-free
2 generic bar code labels per plate
Clear
96
200 μL
Reverse transcription (RT)-PCR
50/Cs.
50/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Thermowell™ 96-Well Polypropylene PCR Microplates > yes; Half Skirt; 96-well; 200 μL