Learn More
Corning™ Costar™ Cell Culture Triangular Flasks
Manufactured from optically clear virgin polystyrene for optimum visibility. Corning™ Cell Culture Triangular Flasks provide excellent accessibility to cell surface.
Brand: Corning™ 3055
This item is not returnable.
View return policy
Description
- Medical-grade polystyrene is sterile and nonpyrogenic
- Tissue culture-treated to promote optimal cell attachment and growth
- Patented canted-neck design improves accessibility for cell monolayer and overall improved handling
- Graduated
- Frosted labeling surfaces
- Extra-wide neck openings for easier pipetting
- Printed lot number and catalog number on each flask
- Vented filter caps
- Maximize gas exchange while minimizing the possibility of contamination
- Hydrophobic 0.2 μm pore size membrane ensures consistent gas exchange without requiring the cap to be loosened
Specifications
Black | |
Polystyrene | |
Angled | |
Tissue Culture Treated | |
Sterile |
HDPE | |
For optimal cell attachment and growth | |
Triangular | |
Yes |
Product Content Correction
Product Title
Corning™ Costar™ Cell Culture Triangular Flasks > 25cm2 Growth area; Phenolic Cap
Spot an opportunity for improvement?Share a Content Correction