Learn More
Corning™ HTS Transwell™ Clear Untreated Reservoirs
Manufactured from clear plastic for maximum visibility. Corning™ HTS Transwell™ Clear Untreated Reservoir is for use with HTS Transwell Permeable Supports.
Brand: Corning™ 3395
This item is not returnable.
View return policy
Description
- For use as reservoir tray with HTS Transwell Permeable Supports
- Not Treated for cell attachment
- Same footprint as standard 24 well plate but with single rectangular well
Specifications
Clear | |
Sterile | |
Cell Culture Insert System | |
48/Cs. |
Polystyrene | |
Non-treated | |
HTS Transwell™ Permeable Supports | |
12/Pk. |
Product Content Correction
Product Title
Corning™ HTS Transwell™ Clear Untreated Reservoirs > Microplates; HTS Transwell Open Reservoir; 48/Cs.
Spot an opportunity for improvement?Share a Content Correction