missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cryo Aprons

Protect your clothing with Thermo Scientific™ Cryo Aprons, available in small, medium and large sizes.

563.81 € - 626.12 €

Specifications

Type Cryogenic Apron
Color Blue
View More Specs

Products 3
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
16449019
View Documents
Thermo Scientific™
189451
Small
617.07 €
Each
Estimated Shipment: 08-04-2025
to see stock.
Add to basket
 
15425764
View Documents
Thermo Scientific™
189453
Large
626.12 €
Each
In Stock
to see stock.
Edge
Add to basket
 
11766287
View Documents
Thermo Scientific™
189452
Medium
563.81 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Protect your clothing with Thermo Scientific™ Cryo Aprons, available in small, medium and large sizes.

  • Small: 36 in. (91.4cm)
  • Medium: 42 in. (106.7cm)
  • Large: 48 in. (121.9cm)
  • Warranty varies by country, please contact us for details
Specifications

Specifications

Cryogenic Apron
Blue
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.