Learn More
Thermo Scientific Chemicals Crystal Violet, certified
Brand: Thermo Scientific Chemicals 405830250
31.30 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Crystal Violet | |
0.98 to 1.2 (P-15/P+15) | |
C25H30ClN3 | |
13, 756 | |
Basic Violet 3, C.I. 42555, Gentian Violet | |
Authentic | |
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride | |
11057 | |
407.99 | |
Green | |
Crystalline Powder |
548-62-9, 90-94-8 | |
88% min. | |
MFCD00011750 | |
15, 4430 | |
ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] | |
407.986 | |
CHEBI:41688 | |
Certified | |
25 g |
Safety and Handling
- Crystal Violet
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H350-May cause cancer.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : C.I. Basic violet 1,Michler's ketone
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468475 | Certificate of Analysis | 31/01/2025 | 405830250,405831000,405835000 |
A0465674 | Certificate of Analysis | 26/08/2024 | 405830250,405831000,405835000 |
B0149353 | Certificate of Analysis | 14/11/2023 | 405830250,405831000,405835000 |
A0457619 | Certificate of Analysis | 09/11/2023 | 405830250,405831000,405835000 |
B0148765 | Certificate of Analysis | 20/10/2023 | 405830250,405831000,405835000 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only