Learn More
Intelsius™ PathoShield™
The PathoShield™ is a complete shipping solution designed for the transportation of Category A and Category B biological materials. Each system is pre-printed with the required markings and includes all the components required to transport your sample safely.
Brand:  Intelsius™ PA025
Encompass_Preferred
 View more versions of this product
Product Code. 15681632
  • 195.45 € / Pack of 25
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Complete Category B shipping solution includes flexible 95kPa tested compliant secondary, absorbent, labels, and relevant UN markings
  • Range of sizes suitable for shipping through postal and courier networks
  • Suitable for a wide range of primaries including blood vials, swabs and sample containers
  • Supplied flat packed for efficient storage and shipping prior to use
Specifications
Specifications
Pouch
Sample Transport Packaging
275 x 179 x 43 mm (10.8 x 7.0 x 1.7 in.)
5
Pouch: 150 x 190 mm (5.9 x 7.4 in.)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Intelsius™ PathoShield™