missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Glass Erlenmeyer Cell Culture Flask with Fluted Neck
Culture your tissues using the Fisherbrand™ Glass Erlenmeyer Cell Culture Flask with Fluted Neck. It is designed for consistency, cell health and reproducability with surface and ancillary options you're looking for in a issue culture flask.
40.15 € - 73.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11532283
|
Fisherbrand™
FB55901 |
250 mL |
40.15 €
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
11542283
|
Fisherbrand™
FB55903 |
500 mL |
49.30 €
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
11552283
|
Fisherbrand™
FB55905 |
1000 mL |
60.25 €
Each |
Estimated Shipment: 14-04-2025
Log in to see stock. |
||||||
11562283
|
Fisherbrand™
FB55907 |
2000 mL |
73.00 €
Each |
In Stock
Log in to see stock. |
||||||
Description
- Fluted neck
- For adherent or suspension cells
- 1/3 turn cap enables one-handed operation
Specifications
Borosilicate Glass | |
Fluted | |
Conical |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Glass Erlenmeyer Cell Culture Flask with Fluted Neck