Learn More
Fisherbrand™ Glass Erlenmeyer Cell Culture Flask with Fluted Neck
Culture your tissues using the Fisherbrand™ Glass Erlenmeyer Cell Culture Flask with Fluted Neck. It is designed for consistency, cell health and reproducability with surface and ancillary options you're looking for in a issue culture flask.
Brand:  Fisherbrand™ FB55903
 View more versions of this product
Product Code. 11542283
  • 49.30 € / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Fluted neck
  • For adherent or suspension cells
  • 1/3 turn cap enables one-handed operation
Specifications
Specifications
500 mL
Borosilicate Glass
Fluted
Conical
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Glass Erlenmeyer Cell Culture Flask with Fluted Neck > Capacity: 500mL; Outer Diameter: 105mm