Learn More
IKA™ MF 10.1 Impact Grinding Head
For crushing fibrous substances such as paper and vegetation, but also for plastics and material with a low volume weight.
Brand:  IKA™ 0002870900
Product Code. 10791941
  • 4465.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Before being discharged, the ground material passes through a sieve. This sieve is interchangeable and available in different hole sizes (not incl. with delivery). The ground material can then be collected using an NS 29 standard ground vessel.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cutting-grinding Head
For Plastics and Material with a Low Volume Weight
Stainless Steel
2.2 kg
320 mm
300 x 320 x 560 mm
560 mm
24.5 m/sec.
12.6 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
IKA™ MF 10.1 Impact Grinding Head > Speed: 24.5m/sec