Learn More
Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips)
GREENER_CHOICE
Achieve ultimate performance in automated pipetting with these tips--superior quality with the flexibility to choose tips for any application.
246.00 € - 2875.00 €
Products 22
Product Code Brand Volume (Metric) Sterility Filtered Packaging Price Quantity & Availability  
Product Code Brand Volume (Metric) Sterility Filtered Packaging Price Quantity & Availability  
10089982
GREENER_CHOICE
View Documents
Thermo Scientific™
5301
12.5 μL Non-sterile Non-filtered 10 x 384 tips
1030.00 €
Pack of 3840
Estimated Shipment: 11-09-2025
to see stock.
Add to basket
10794434
GREENER_CHOICE
View Documents
Thermo Scientific™
5302
12.5 μL Sterile Non-filtered 10 x 384 tips
1050.00 €
Pack of 3840
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
10592285
GREENER_CHOICE
View Documents
Thermo Scientific™
5317
30 μL Sterile Non-filtered 20 x 384 tips
2015.00 €
Pack of 7680
In Stock
to see stock.
Add to basket
10502295
View Documents
Thermo Scientific™
5318
30 μL Sterile Filtered 20 x 384 tips
2875.00 €
Pack of 7680
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
10283293
GREENER_CHOICE
View Documents
Thermo Scientific™
5321
100 μL Non-sterile Non-filtered 10 x 384 tips
1030.00 €
Pack of 3840
Estimated Shipment: 11-09-2025
to see stock.
Add to basket
12987197
GREENER_CHOICE
View Documents
Thermo Scientific™
5326
100 μL Non-sterile Non-filtered 20 x 384 tips
1985.00 €
Pack of 7680
In Stock
to see stock.
Add to basket
11719048
GREENER_CHOICE
View Documents
Thermo Scientific™
5327
100 μL Sterile Non-filtered 20 x 384 tips
2015.00 €
Pack of 7680
In Stock
to see stock.
Add to basket
10203443
GREENER_CHOICE
View Documents
Thermo Scientific™
5328
100 μL Sterile Filtered 20 x 384 tips
2875.00 €
Pack of 7680
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
10573625
GREENER_CHOICE
View Documents
Thermo Scientific™
5526
100 μL Non-sterile Non-filtered 20 x 96 tips
397.00 €
Pack of 1920
Estimated Shipment: 11-09-2025
to see stock.
Add to basket
11567459
GREENER_CHOICE
View Documents
Thermo Scientific™
5527
100 μL Sterile Non-filtered 20 x 96 tips
418.00 €
Pack of 1920
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
11577459
GREENER_CHOICE
View Documents
Thermo Scientific™
5528
100 μL Sterile Filtered 20 x 96 tips
627.00 €
Pack of 1920
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
11507479
GREENER_CHOICE
View Documents
Thermo Scientific™
5586
30 μL Non-sterile Non-filtered 20 x 96 tips
358.00 €
Pack of 1920
In Stock
to see stock.
Add to basket
10572285
GREENER_CHOICE
View Documents
Thermo Scientific™
5587
30 μL Sterile Non-filtered 20 x 96 tips
379.00 €
Pack of 1920
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
10532865
GREENER_CHOICE
View Documents
Thermo Scientific™
5588
30 μL Sterile Filtered 20 x 96 tips
622.00 €
Pack of 1920
Estimated Shipment: 09-09-2025
to see stock.
Add to basket
10541715
GREENER_CHOICE
View Documents
Thermo Scientific™
5311
30 μL Non-sterile Non-filtered 10 x 384 tips
1030.00 €
Pack of 3840
Estimated Shipment: 11-09-2025
to see stock.
Add to basket
10485953
View Documents
Thermo Scientific™
5541
220 μL Non-sterile Non-filtered 20 x 96 tips
246.00 €
Pack of 1920
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
10495953
GREENER_CHOICE
View Documents
Thermo Scientific™
5542
250 μL Sterile Non-filtered 20 x 96 tips
268.00 €
Pack of 1920
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
10101633
GREENER_CHOICE
View Documents
Thermo Scientific™
5545
220 μL Sterile Filtered 20 x 96 tips
426.00 €
Pack of 1920
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
10747314
View Documents
Thermo Scientific™
5571
30 μL Non-sterile Non-filtered 20 x 96 tips
277.00 €
Pack of 1920
Estimated Shipment: 11-09-2025
to see stock.
Add to basket
10171393
GREENER_CHOICE
View Documents
Thermo Scientific™
5322
100 μL Sterile 10 x 384 tips
1055.00 €
Pack of 3840
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
12917207
GREENER_CHOICE
View Documents
Thermo Scientific™
5572
30 μL Sterile Non-filtered 20 x 96 tips
308.00 €
Pack of 1920
Estimated Shipment: 18-09-2025
to see stock.
Add to basket
10355073
GREENER_CHOICE
View Documents
Thermo Scientific™
5316
30 μL Non-sterile Non-filtered 20 x 384 tips
2005.00 €
Pack of 7680
In Stock
to see stock.
Add to basket
Description
Description
Perform pipetting more effectively with Thermo Scientific™ Matrix™ SerialMate™ Pipette Tips, for use with the Matrix SerialMate™ Pipetter. Unlike conventional automation tips, D.A.R.T.'s™ are packaged in disposable rigid magazines and seal onto the pipetting heads by being pressed against a silicone pad. This definitive seal minimizes volume displacement, ensures tip straightness and enables consistent dispensing for improved well access. A small internal orifice provides small droplet formation resulting in optimal accuracy and precision.
Features:
  • Stepped-body design minimizes volume displacement and ensures tip straightness for better well access.
  • Highly polished molds and cores ensure low liquid retention.
  • Small internal tip orifice provides better small volume droplet formation
  • Sterile, sterile filtered, wide-bore, and extended-length tip options are available
  • Tips are thoroughly inspected for proper formation and straightness
  • Unique face seal design ensures consistent tip-to-tip height for precise low volume dispensing.
  • Entire assembly can be quickly removed and replaced eliminating extra handling of the tips, which can cause damage or contamination.
Includes: 20 magazines per case
Compatible with: Thermo Scientific™ Matrix™ Hydra™ DT, Matrix™ PlateMate™ 2x3 and PlateMate™ Plus Systems.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ D.A.R.T.'S™ (Disposable Automation Research Tips)