missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-(-)-Asparagine monohydrate, 99% Product Code.: 11400398

Thermo Scientific Chemicals D-(-)-Asparagine monohydrate, 99%

Product Code. 11400398
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11400398

Brand: Thermo Scientific Chemicals B24556.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-(-)-Asparagine monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Asparagine monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water (35 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5794-24-1
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.134
MDL Number MFCD00149558
InChI Key RBMGJIZCEWRQES-HSHFZTNMSA-N
Synonym d-asparagine monohydrate, d-asparagine hydrate, r-2,4-diamino-4-oxobutanoic acid hydrate, d-asparagine, monohydrate, d-asparaginemonohydrate, d---asparagine monohydrate, d-aspartic acid 4-amide, r---2-aminosuccinamic acid, d--asparagine monohydrate, r-2-aminosuccinic acid 4-amideShow MoreShow Less
PubChem CID 16211197
IUPAC Name (2R)-2,4-diamino-4-oxobutanoic acid;hydrate
SMILES C(C(C(=O)O)N)C(=O)N.O

Specifications

Melting Point 274°C to 276°C (decomposition)
Merck Index 14,837
Solubility Information Soluble in water (35g/L at 20°C).
Quantity 25 g
Formula Weight 150.14 (132.12 Anhydrous)
Percent Purity 99%
Chemical Name or Material D-(-)-Asparagine monohydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10115317 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog NumberB24556.22,B24556.14
Lot Number10243284 Certificate TypeCertificate of Analysis Date16/09/2022 Catalog NumberB24556.22,B24556.14
Lot Number10115317 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24556.22,B24556.14
Lot Number10125841 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24556.22,B24556.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-(-)-Asparagine monohydrate, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.