missing translation for 'onlineSavingsMsg'
Learn More
D-beta-Proline, 98+%, Thermo Scientific Chemicals Product Code.: 15403848

D-beta-Proline, 98+%, Thermo Scientific Chemicals

Product Code. 15403848
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15403848

Brand: Thermo Scientific Alfa Aesar H57356.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72580-54-2
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD08272856
InChI Key JAEIBKXSIXOLOL-SCSAIBSYSA-N
Synonym r-pyrrolidine-3-carboxylic acid, 3r-pyrrolidine-3-carboxylic acid, r---pyrrolidine-3-carboxylic acid, d-beta-proline, r-beta-proline, 3-pyrrolidinecarboxylic acid, 3r, r-3-pyrrolidinecarboxylic acid, l-pyrrolidine-3-carboxylic acid, 3r-3-pyrrolidinecarboxylic acid hcl, r-pyrrilidine-3-carboxylic acidShow MoreShow Less
PubChem CID 1501970
IUPAC Name (3R)-pyrrolidine-3-carboxylic acid
SMILES C1CNCC1C(=O)O

Specifications

Melting Point 185°C to 191°C
Beilstein 5496144
Quantity 250 mg
Formula Weight 115.13
Optical Rotation −18° (c=2 in Water)
Percent Purity ≥98%
Chemical Name or Material D-beta-Proline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberK12Y020 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog NumberH57356.MD,H57356.03
Lot NumberY10G041 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57356.MD,H57356.03
Lot NumberS19F004 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57356.MD,H57356.03
Lot NumberK12Y020 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57356.MD,H57356.03
Lot NumberZ04F018 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57356.MD,H57356.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-beta-Proline, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.