missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-Leucine, 99% Product Code.: 10357980

Thermo Scientific Chemicals D-Leucine, 99%

Product Code. 10357980
10 g, Glass Bottle
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10357980

Brand: Thermo Scientific Chemicals 226900100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-38-1
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00063088
InChI Key ROHFNLRQFUQHCH-RXMQYKEDSA-N
Synonym d-leucine, h-d-leu-oh, r-leucine, 2r-2-amino-4-methylpentanoic acid, d-homo-valine, r---leucine, leucine, d, r-2-amino-4-methylpentanoic acid, d-leucin, d-leuzinShow MoreShow Less
PubChem CID 439524
ChEBI CHEBI:28225
IUPAC Name (2R)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@@H](N)C(O)=O

Specifications

Arsenic (As) 2ppm max.
Color White
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99%
Loss on Drying 0.2% max.
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2CH(NH2)CO2H
Residue after Ignition 0.1% max.
Beilstein 04, 446
Specific Rotation -14.9° to -16° (20°C, 589nm) (c=4, 6N HCl)
Specific Rotation Condition −15.45° (20°C c=4,6N HCl)
Quantity 10 g
Formula Weight 131.17
Percent Purity ≥98.5%
Physical Form Crystalline Powder, Crystals, or Flakes
Chemical Name or Material D-Leucine, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0456041 Certificate TypeCertificate of Analysis Date01/09/2023 Catalog Number226900100,226900025
Lot NumberA0450729 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number226900100,226900025
Lot NumberA0355501 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number226900100,226900025
Lot NumberA0441209 Certificate TypeCertificate of Analysis Date27/04/2022 Catalog Number226900100,226900025
Lot NumberA0429013 Certificate TypeCertificate of Analysis Date03/09/2021 Catalog Number226900100,226900025
Product Identifier
  • D-Leucine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-Leucine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.