missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Leucine, 99% Product Code.: 10274980

Thermo Scientific Chemicals L-Leucine, 99%

Product Code. 10274980
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 10274980

Brand: Thermo Scientific Chemicals 125121000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Leucine, CAS # 61-90-5, is a branched-chain aliphatic essential amino acid in humans that is important for protein synthesis and many metabolic functions.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L-Leucine is an essential amino acid that has an important role in the growth and repair of muscle and bone tissue
  • It functions to preserve muscle glycogen and helps to prevent the breakdown of muscle proteins after trauma or severe stress
  • It is also involved in hemoglobin formation and metabolic functions. This amino acid cannot be synthesized to meet the body's needs from simpler molecules, and therefore must be provided in the diet.
  • L-Leucine stimulates protein synthesis by the initiation of mRNA translation

Applications

  • L-Leucine is used in the biosynthesis of proteins
  • In vivo studies show that L-Leucine can be used when gene defects lead to an organism's inability to metabolize the amino acid phenylalanine
  • This compound also contributes to the regulation of blood-sugar levels, growth hormone production, and wound healing
  • In research experiments this compound has been used as an activator of the mammalian target of rapamycin (mTOR)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acidShow MoreShow Less
PubChem CID 6106
ChEBI CHEBI:15603
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

Melting Point 286.0°C to 288.0°C
Specific Rotation + 15.40
Solubility Information Solubility in water: 22.4g/L (20°C). Other solubilities: 10.9g/L acetic acid, slightly soluble in alcohol, insoluble in ether
Sublimation Point 145.0 °C
Specific Rotation Condition + 15.40 (20.00°C c=4, 6N HCl)
Quantity 100 g
Formula Weight 131.17
Chemical Name or Material L-Leucine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB0154401 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number125125000,125121000,125120025
Lot NumberB0150359 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number125125000,125121000,125120025
Lot NumberB0150483 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number125125000,125121000,125120025
Lot NumberB0149340 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number125125000,125121000,125120025
Lot NumberB0147174 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number125125000,125121000,125120025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Leucine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.