missing translation for 'onlineSavingsMsg'
Learn More
D-Serine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals Product Code.: 15470499

D-Serine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15470499
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15470499

Brand: Thermo Scientific Alfa Aesar H63685.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-Serine methyl ester hydrochloride is used as a intermediate for organic synthesis, pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Serine methyl ester hydrochloride is used as a intermediate for organic synthesis, pharmaceutical and chemical research.

Solubility
Soluble in ethanol and methanol.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5874-57-7
Molecular Formula C23H19Br2N3O3S
Molecular Weight (g/mol) 577.29
MDL Number MFCD00066121
InChI Key LYRWWUCOZBTOPL-UHFFFAOYNA-N
Synonym d-serine methyl ester hydrochloride, h-d-ser-ome.hcl, r-methyl 2-amino-3-hydroxypropanoate hydrochloride, d-serine, methyl ester, hydrochloride, d-serine methyl ester hcl, methyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-ser-ome hcl, cycloserine impurity 2, h-d-ser-ome.hciShow MoreShow Less
PubChem CID 11446470
SMILES CC(N(C)S(=O)(=O)C1=CC=C(Br)C=C1)C1=NC2=CC=CC=C2C(=O)N1C1=CC=C(Br)C=C1

Specifications

Melting Point 163°C to 166°C (decomposition)
Solubility Information Soluble in ethanol and methanol.
IUPAC Name methyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride
Quantity 5 g
Formula Weight 155.58
Percent Purity 95%
Physical Form Solid
Chemical Name or Material D-Serine methyl ester hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU10J010 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberH63685.22,H63685.14,H63685.06
Lot NumberT27I002 Certificate TypeCertificate of Analysis Date04/08/2022 Catalog NumberH63685.22,H63685.14,H63685.06
Lot NumberR17I017 Certificate TypeCertificate of Analysis Date06/06/2022 Catalog NumberH63685.22,H63685.14,H63685.06
Lot Number10238290 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH63685.22,H63685.14,H63685.06
Lot Number81301657 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberH63685.22,H63685.14,H63685.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-Serine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.