missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-Threonine, 98% Product Code.: 10076233

Thermo Scientific Chemicals D-Threonine, 98%

Product Code. 10076233
100 g, Glass Bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10076233

Brand: Thermo Scientific Chemicals 346971000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-20-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064269
InChI Key AYFVYJQAPQTCCC-STHAYSLISA-N
Synonym d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dthShow MoreShow Less
PubChem CID 69435
ChEBI CHEBI:16398
IUPAC Name (2R,3S)-2-amino-3-hydroxybutanoic acid
SMILES C[C@H](O)[C@@H](N)C(O)=O

Specifications

Ammonium 200ppm max.
Arsenic (As) 1ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 98%
Loss on Drying 0.3% max.
Other Amino Acids (TLC) Passes Test
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH(NH)COH
Specific Rotation +27° to +29° (20°C, 589nm) (c=6, water)
Sulfate 300ppm max.
Transmittance 98% min. (c=5, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Specific Rotation Condition +28° (20°C c=6,water)
Quantity 100 g
Formula Weight 119.12
Percent Purity ≥97.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D-Threonine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0417508 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number346971000,346970250,346970050
Lot NumberA0447287 Certificate TypeCertificate of Analysis Date04/11/2022 Catalog Number346971000,346970250,346970050
Lot NumberA0382859 Certificate TypeCertificate of Analysis Date04/04/2022 Catalog Number346971000,346970250,346970050
Lot NumberA0432427 Certificate TypeCertificate of Analysis Date26/07/2021 Catalog Number346971000,346970250,346970050
Product Identifier
  • D-Threonine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-Threonine, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.