missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-Threonine, 99% Product Code.: 11484667

Thermo Scientific Chemicals D-Threonine, 99%

Product Code. 11484667
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11484667

Brand: Thermo Scientific Chemicals B21177.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-20-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064269
InChI Key AYFVYJQAPQTCCC-STHAYSLISA-N
Synonym d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dthShow MoreShow Less
PubChem CID 69435
ChEBI CHEBI:16398
IUPAC Name (2R,3S)-2-amino-3-hydroxybutanoic acid
SMILES C[C@H](O)[C@@H](N)C(O)=O

Specifications

Melting Point ∼274°C (decomposition)
Beilstein 1721643
Quantity 5 g
Formula Weight 119.12
Optical Rotation +29° (c=5 in Water)
Percent Purity 99%
Chemical Name or Material D-Threonine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251017 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB21177.22,B21177.06,B21177.14
Lot Number10244097 Certificate TypeCertificate of Analysis Date20/10/2022 Catalog NumberB21177.22,B21177.06,B21177.14
Lot NumberT06I012 Certificate TypeCertificate of Analysis Date19/07/2022 Catalog NumberB21177.22,B21177.06,B21177.14
Lot Number5037A12W Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21177.22,B21177.06,B21177.14
Lot Number10225939 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21177.22,B21177.06,B21177.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-Threonine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.