missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-Valine, 98+% Product Code.: 11453138

Thermo Scientific Chemicals D-Valine, 98+%

Product Code. 11453138
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11453138

Brand: Thermo Scientific Chemicals A18894.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 640-68-6
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064219
InChI Key KZSNJWFQEVHDMF-SCSAIBSYSA-N
Synonym d-valine, h-d-val-oh, r-valine, valine, d, d-val, 2r-2-amino-3-methylbutanoic acid, d-2-aminoisovaleric acid, d-valin, unii-y14i1443ur, d-2-amino-3-methylbutanoic acidShow MoreShow Less
PubChem CID 71563
ChEBI CHEBI:27477
IUPAC Name (2R)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@@H](N)C(O)=O

Specifications

Melting Point 295°C (decomposition)
Beilstein 1721135
Quantity 5 g
Formula Weight 117.15
Optical Rotation −27° (c=5 in 5N HCl)
Percent Purity ≥98%
Chemical Name or Material D-Valine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10206455 Certificate TypeCertificate of Analysis Date13/07/2023 Catalog NumberA18894.22,A18894.14,A18894.06
Lot Number10134416 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18894.22,A18894.14,A18894.06
Lot Number10132310 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18894.22,A18894.14,A18894.06
Lot Number10142378 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18894.22,A18894.14,A18894.06
Lot Number10150416 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18894.22,A18894.14,A18894.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-Valine, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.