Learn More
Corning™ DeckWorks™ Reload Systems Starter Kits
Contains one DeckWorks Reload System with standard tips and empty hinged racks
Brand: Corning™ 4133
This item is not returnable.
View return policy
Description
DeckWorks starter kits contain one DeckWorks reload system with standard tips and empty hinged racks.
- 10 empty racks accompany 10 μL, 200 μL, and 300 μL volume tips.
- 8 empty racks accompany 1000 μL volume tips.
Specifications
1 to 300 μL | |
Natural | |
Refill | |
Starter kit | |
DeckWorks |
Non-sterile | |
Designed to allow to maximize storage space and help reduce the production and waste of plastic materials | |
Round | |
Yes | |
Polypropylene |
Product Suggestions
Customers who viewed this item also viewed
COR008 DeckWorks Pipet Tips
File size: 2.6 MB
COR012 LIQUID HANDLING SELECTION GUIDE
File size: 2.5 MB
Product Content Correction
Product Title
Corning™ DeckWorks™ Reload Systems Starter Kits > Natural; 1-300 μL
Spot an opportunity for improvement?Share a Content Correction