missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyltin dichloride, 96%, Thermo Scientific Chemicals

Product Code. 11367717
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Organotin intermediate

Organotin intermediate.Di-n-butyltin dichloride is used as an organotin intermediate. It serves as heat and light stabilizer for polyvinyl chloride plastics. It is also used in the preparation of methyl 2-(methoxycarbonylmethyl)-5-iodobenzene by reacting with 2,5-diiodobenzoic acid. It acts as an esterification catalyst and a veterinary vermicide and tapeworm remedy. Further, it is used as ion exchange agent, process regulators and processing aids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 683-18-1
Molecular Formula C8H18Cl2Sn
Molecular Weight (g/mol) 303.842
MDL Number MFCD00000518
InChI Key RJGHQTVXGKYATR-UHFFFAOYSA-L
Synonym dibutyltin dichloride, dibutyldichlorotin, di-n-butyltin dichloride, stannane, dibutyldichloro, dibutyltin chloride, dichlorodibutyltin, dibutyl dichloro stannane, dichlorodibutylstannane, dibutyltindichloride, dibutyl tin dichloride
PubChem CID 12688
IUPAC Name dibutyl(dichloro)stannane
SMILES CCCC[Sn](CCCC)(Cl)Cl

Specifications

Melting Point 39°C to 41°C
Color White
Density 1.36 g/mL
Boiling Point 135°C (10 mmHg)
Flash Point 168°C (334°F)
Odor Acrid
Linear Formula [CH3(CH2)3]2SnCl2
Refractive Index 1.4991
Quantity 1 kg
UN Number UN2928
Sensitivity Moisture sensitive
Solubility Information Insoluble in cold water; Hydrolyzed by hot water; Soluble in organic solvents;Insoluble in cold water; Soluble in benzene,toluene,ether and alcohol.
Formula Weight 303.83
Percent Purity 96%
Physical Form Crystalline
Chemical Name or Material Di-n-butyltin dichloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'