missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Fisher Scientific™ Digital Heavy Duty Vortex Mixer

Designed for continuous duty. Fisherbrand Digital Heavy Duty Vortex Mixer features heavy-duty design enables accessories to run throughout the speed range.

Brand:  Fisherbrand™ 80863027

Product Code. 15569014

  • 1090.00 € / Each
Estimated Shipment: 30-07-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • LED displays and touchpad controls for speed and time.
  • Euro style cord with UK and CH adapter
  • Optional accessories including foam inserts for variety of tubes and microtubes, microplate holders, and holders for flasks
  • Threaded accessory attachment and robust motor enable samples to be run through out speed range
TRUSTED_SUSTAINABILITY
Specifications

Specifications

4.9 mm
On mode: 300 to 2500 rpm, Touch mode: 300 to 3500 rpm
Vortex Mixer
1.1 kg
230 V
Universal holder for holding foam tube inserts, universal cover, foam insert for (38) 1,5 - 2mL microtubes, and cuphead
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Fisher Scientific™ Digital Heavy Duty Vortex Mixer > Heavy Duty Vortex Digital

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.