missing translation for 'onlineSavingsMsg'
Learn More
Dihydroergotamine methanesulfonate, Thermo Scientific Chemicals Product Code.: 15485999

Dihydroergotamine methanesulfonate, Thermo Scientific Chemicals

Product Code. 15485999
100 mg
Click to view available options
Quantity:
100 mg
1 g
This item is not returnable. View return policy

Product Code. 15485999

Brand: Thermo Scientific Alfa Aesar J63840.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Agonist at vascular serotonin receptors; partial agonist at alpha-adrenergic and dopamine D2 receptors

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Agonist at vascular serotonin receptors
  • Partial agonist at alpha-adrenergic and dopamine D2 receptors
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6190-39-2
Molecular Formula C34H41N5O8S
Molecular Weight (g/mol) 679.79
MDL Number MFCD00058615
InChI Key ADYPXRFPBQGGAH-UMYZUSPBSA-N
Synonym dihydroergotamine mesylate, dihydroergotamine methanesulfonate salt, dihydroergotoxine, prestwick_746, dihydroergotamine methanesulfonate salt, powder, dihydroergotamine mesilate, european pharmacopoeia ep reference standard, dihydroergotamine for peak identification, european pharmacopoeia ep reference standard, dihydroergotamine mesylate, united states pharmacopeia usp reference standard, 4r,7r-n-1s,2s,4r,7s-7-benzyl-2-hydroxy-4-methyl-5,8-dioxo-3-oxa-6,9-diazatricyclo 7.3.0.0^ 2,6 dodecan-4-yl-6-methyl-6,11-diazatetracyclo 7.6.1.0^ 2,7 .0^ 12,16 hexadeca-1 15 ,9,12 16 ,13-tetraene-4-carboxamide; methanesulfonic acid, 6ar,9r-n-2r,5s,10as,10bs-5-benzyl-10b-hydroxy-2-methyl-3,6-dioxooctahydro-8h-oxazolo 3,2-a pyrrolo 2,1-c pyrazin-2-yl-7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo 4,3-fg quinoline-9-carboxamide methanesulfonateShow MoreShow Less
PubChem CID 6420006
IUPAC Name (2R,4R,7R)-N-[(1S,2S,4R,7S)-7-benzyl-2-hydroxy-4-methyl-5,8-dioxo-3-oxa-6,9-diazatricyclo[7.3.0.0²,⁶]dodecan-4-yl]-6-methyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraene-4-carboxamide; methanesulfonic acidShow MoreShow Less
SMILES CS(O)(=O)=O.CN1C[C@@H](C[C@H]2[C@H]1CC1=CNC3=CC=CC2=C13)C(=O)N[C@]1(C)O[C@@]2(O)[C@@H]3CCCN3C(=O)[C@H](CC3=CC=CC=C3)N2C1=OShow MoreShow Less

Specifications

Melting Point 230°C to 235°C
Color Pink to White
Quantity 100 mg
Merck Index 14,3174
Formula Weight 679.78
Physical Form Powder
Chemical Name or Material Dihydroergotamine methanesulfonate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ20K067 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberJ63840.03,J63840.MC
Lot NumberZ17K009 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberJ63840.03,J63840.MC
Lot NumberT18I026 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog NumberJ63840.03,J63840.MC
Lot NumberR03I047 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog NumberJ63840.03,J63840.MC
Lot NumberG20Y042 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ63840.03,J63840.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dihydroergotamine methanesulfonate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.