missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyltin dichloride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 103.0°C to 110.0°C |
Color | White to Yellow |
Boiling Point | 188.0°C to 190.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (Argentometry) |
Packaging | Glass bottle |
Molecular Formula | C2H6Cl2Sn |
Linear Formula | (CH3)2Sn(Cl)2 |
Quantity | 100 g |
Solubility Information | Solubility in water: 20g/L (20°C). Other solubilities: 0.1 g/mL in methanol |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Safety and Handling



Product Identifier
- Dimethyltin dichloride
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 1
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H361d-Suspected of damaging the unborn child.
- H311-Toxic in contact with skin.
- H301-Toxic if swallowed.
- EUH071-Corrosive to the respiratory tract.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dimethyltin dichloride, 95%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction