Learn More
Lovibond™ Comparator Disc
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
Brand: Lovibond™ 234020
Description
Lovibond™ Comparator Disc uses an array of glass fade-free color match standards for chemical test color comparison.
- Easy to use, visual assessment of the sample; simply rotate disc to match color
- Glass, fade-free standards for longevity of use
- Shock resistant, scratch resistant
- Optional lighting units for guaranteed lighting conditions 24 hours/day
- Optional Nessleriser systems for longer path lengths
- More than 400 color discs are available for the Comparator 2000+
- Lovibond™ color standards consist exclusively of through-and-through colored and flat-ground glasses
Specifications
Chlorine | |
Comparator 2000+ | |
Color Comparator Disc |
Disc 3/40B | |
0.2, 0.4, 0.6, 1.0, 1.5, 2.0, 2.5, 3.0, 4.0mg/L |
Product Suggestions
Customers who viewed this item also viewed.