Learn More
Buerkle™ Disposable Funnel for Powders, LaboPlast™/SteriPlast™
BIOPHARMAGREENER_CHOICE
The Disposable Funnel for Powders, LaboPlast™/SteriPlast™ has a short, wide discharge and is suitable for transferring powdered samples and materials, granulates, and suspensions, as well as other liquids.
Brand:  Buerkle™ 5378-6007
 View more versions of this product
Product Code. 17540846
  • 32.80 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For easy and reliable transferring
  • Wide opening prevents clogging
  • Suitable for use in laboratories and clean rooms or for sampling
  • Clean room manufactured (clean room class 7)
  • Individually packaged and optionally sterilized
  • Complies with EU food requirements and FDA regulations
Specifications
Specifications
100 mm
Polystyrene
Transferring Powders, Granulates and Suspensions
Filling Funnel (Powders)
25 mm
Blue
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ Disposable Funnel for Powders, LaboPlast™/SteriPlast™ > Color: Blue; Material: PS; Sterility: Sterile