Learn More
Brady™ Dissolvable Paper Labels for Thermal Transfer Printers
GREENER_CHOICE
These water dissolvable labels can be washed away without leaving behind any residue.
Brand:  Brady™ 198764
 View more versions of this product
Product Code. 15965760
  • 118.00 € / Pack of 1000
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The water dissolvable paper label is self-adhesive and extremely easy to apply. Exposing the label to warm water will completely dissolve it within 30 seconds, without damage to the surface it was applied to.
  • Leaves no residue
  • Reduces labor required to remove labels
  • Reduces maintenance to filters and drains
  • Enables easy reidentification of reusable items in a range of industries
Specifications
Specifications
Not Autoclavable
Acrylic
Matte
Thermal Transfer
Asset Tracking, General Identification, Laboratory Labeling, Warehouse Marking
1
Rectangle
0.076 mm
101.6 mm
THT-55-403-1
White
Paper (B-403)
Non-sterile
BradyPrinter i5100, i7100, and PR Plus Series Label Printers, IP™ Series Printer, THT 76 mm Core Printers
1000 Labels
-70°C to +100°C
54 mm
50.8 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ Dissolvable Paper Labels for Thermal Transfer Printers > Dissolvable Paper Labels; Color: White; Height (Metric): 50.8 mm; Labels per Row: 1; Vertical Repeat (Metric): 54 mm; Width (Metric): 101.6 mm