missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Leucine, 99% Product Code.: 11493254

Thermo Scientific Chemicals DL-Leucine, 99%

Product Code. 11493254
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11493254

Brand: Thermo Scientific Chemicals A10590.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-39-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00063087
InChI Key ROHFNLRQFUQHCH-UHFFFAOYNA-N
Synonym dl-leucine, h-dl-leu-oh, leucine, dl, rs-leucine, +--leucine, dl-2-amino-4-methylpentanoic acid, 2-amino-4-methyl-pentanoic acid, l-1-13c leucine, l-1-14c leucine, leucine,dShow MoreShow Less
PubChem CID 857
ChEBI CHEBI:25017
IUPAC Name 2-amino-4-methylpentanoic acid
SMILES CC(C)CC(N)C(O)=O

Specifications

Melting Point ∼294°C (sublimation)
Beilstein 636005
Merck Index 14,5451
Density 1.293
Quantity 250 g
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material DL-Leucine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX25K011 Certificate TypeCertificate of Analysis Date08/11/2024 Catalog NumberA10590.30,A10590.18,A10590.0B
Lot Number10219733 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog NumberA10590.30,A10590.18,A10590.0B
Lot NumberU17J009 Certificate TypeCertificate of Analysis Date06/09/2023 Catalog NumberA10590.30,A10590.18,A10590.0B
Lot NumberS19J056 Certificate TypeCertificate of Analysis Date23/06/2023 Catalog NumberA10590.30,A10590.18,A10590,A10590.0B
Lot NumberZ19I028 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog NumberA10590.30,A10590.18,A10590.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Leucine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.