missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Methionine, 99+% Product Code.: 10457130

Thermo Scientific Chemicals DL-Methionine, 99+%

Product Code. 10457130
100 g, Plastic Bottle
Click to view available options
Quantity:
2.5 g
100 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10457130

Brand: Thermo Scientific Chemicals 125651000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-51-8
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063096
InChI Key FFEARJCKVFRZRR-UHFFFAOYNA-N
Synonym dl-methionine, methionine, racemethionine, acimetion, banthionine, cynaron, lobamine, meonine, mertionin, metione
PubChem CID 876
ChEBI CHEBI:16811
SMILES CSCCC(N)C(O)=O

Specifications

Ammonium 1000ppm max.
pH 5.4 to 6.1 (2% w/v aq. soln.)
Color White
Chloride 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.5% max. (1 g, 105°C)
Other Amino Acids 0.2% max. (TLC)
Packaging Plastic Bottle
Linear Formula CH3SCH2CH2CH(NH2)CO2H
Beilstein 04, II, 938
Sulfate 200ppm max.
Merck Index 15, 6047
Transmittance 95% min. (c=10, 2N HCl, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Density 1.34g/mL
IUPAC Name 2-amino-4-methylsulfanylbutanoic acid
Quantity 100 g
Formula Weight 149.21
Percent Purity 99.0 to 101.0% (on dried substance)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material DL-Methionine, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0427618 Certificate TypeCertificate of Analysis Date17/11/2023 Catalog Number125652500,125650025,125650010,125651000
Lot NumberA0455332 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number125652500,125650025,125650010,125651000
Lot NumberA0402825 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number125652500,125650025,125650010,125651000
Lot NumberA0407950 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number125652500,125650025,125650010,125651000
Lot NumberA0411398 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number125652500,125650025,125650010,125651000
Product Identifier
  • DL-Methionine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Methionine, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.