missing translation for 'onlineSavingsMsg'
Learn More
DL-Serine, ≥98%, MP Biomedicals™ Product Code.: 11466242

DL-Serine, ≥98%, MP Biomedicals™

Product Code. 11466242 Shop All MP Biomedicals Products
25 g
Click to view available options
Quantity:
25 g
1 kg
This item is not returnable. View return policy

Product Code. 11466242

Brand: MP Biomedicals 0210286825

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 302-84-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
InChI Key MTCFGRXMJLQNBG-UHFFFAOYSA-N
Synonym dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid
PubChem CID 617
ChEBI CHEBI:17822
IUPAC Name 2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Color White
Assay Percent Range ≥98%
Density 1.537g/mL
Quantity 25 g
Formula Weight 105.1
Physical Form Crystalline Powder
Chemical Name or Material DL-Serine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberYD02001 Certificate TypeCertificate of Analysis Date24/03/2023 Catalog Number0210286825
Lot NumberSR04643 Certificate TypeCertificate of Analysis Date17/03/2022 Catalog Number0210286825
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-Serine, ≥98%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.