missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Serine, 99% Product Code.: 11482188

Thermo Scientific Chemicals DL-Serine, 99%

Product Code. 11482188
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11482188

Brand: Thermo Scientific Chemicals A15184.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DL-Serine is used in the synthesis of novel tryptoline derivatives as IDO (indoleamine 2,3-Deoxygenase) inhibitors, for potential use in Alzheimer’s treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Serine is used in the synthesis of novel tryptoline derivatives as IDO (indoleamine 2,3-Deoxygenase) inhibitors, for potential use in Alzheimer’s treatment.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-84-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00064223
InChI Key MTCFGRXMJLQNBG-UHFFFAOYSA-N
Synonym dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid
PubChem CID 617
ChEBI CHEBI:17822
IUPAC Name 2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point ∼240°C (decomposition)
Beilstein 1721402
Merck Index 14,8460
Solubility Information Soluble in water.
Quantity 100 g
Formula Weight 105.09
Percent Purity 99%
Chemical Name or Material DL-Serine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10248160 Certificate TypeCertificate of Analysis Date28/09/2023 Catalog NumberA15184.36,A15184.22
Lot Number10238204 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15184.36,A15184.22
Lot Number10125497 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15184.36,A15184.22
Lot NumberG1895A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15184.36,A15184.22
Lot Number10165590 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15184.36,A15184.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Serine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.