missing translation for 'onlineSavingsMsg'
Learn More
DL-Valine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals Product Code.: 11344825

DL-Valine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11344825
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11344825

Brand: Thermo Scientific Alfa Aesar L17906.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23358-42-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00067542
InChI Key PQGVTLQEKCJXKF-UHFFFAOYSA-N
Synonym dl-valine ethyl ester hydrochloride, ethyl 2-amino-3-methylbutanoate hydrochloride, h-dl-val-oet.hcl, ethyl dl-valinate hydrochloride, valine ethyl ester, hydrochloride, ethyl dl-valinate hcl, acmc-20al3t, acmc-209ta5, +/--valine ethyl ester hydrochloride, ethyl 2-azanyl-3-methyl-butanoate hydrochlorideShow MoreShow Less
PubChem CID 13000942
IUPAC Name ethyl 2-amino-3-methylbutanoate;hydrochloride
SMILES Cl.CCOC(=O)C(N)C(C)C

Specifications

Melting Point ∼94°C
Beilstein 3912411
Quantity 25 g
Formula Weight 181.66
Percent Purity 99%
Chemical Name or Material DL-Valine ethyl ester hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10119559 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL17906.06,L17906.14
Lot Number10222367 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL17906.06,L17906.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-Valine ethyl ester hydrochloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.