missing translation for 'onlineSavingsMsg'
Learn More
Dowex™ 1X2, Thermo Scientific Chemicals Product Code.: 10184070

Dowex™ 1X2, Thermo Scientific Chemicals

Product Code. 10184070
1 kg, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10184070

Brand: Thermo Scientific Acros 202950010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9085-42-1
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.05
MDL Number MFCD00132715
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
SMILES [Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1

Specifications

Physical Form Beads
Packaging Plastic bottle
Water Retention Capacity 65 to 75% (Cl form)
Fieser 01,511; 04,267; 06,302; 07,182; 08,263
Quantity 1 kg
Solubility Information Solubility in water: insoluble.
IUPAC Name 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
Chemical Name or Material Dowex™ 1X2

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0326022 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number202950025,202950100,202950010,202951000,202955000
Lot NumberA0326021 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number202950025,202950100,202950010,202951000,202955000
Lot NumberA0409541 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog Number202950025,202950100,202950010,202951000,202955000
Lot NumberA0330016 Certificate TypeCertificate of Analysis Date21/01/2022 Catalog Number202950025,202950100,202950010,202951000,202955000
Lot NumberA0437800 Certificate TypeCertificate of Analysis Date06/01/2022 Catalog Number202950025,202950100,202950010,202951000,202955000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dowex™ 1X2, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.