Learn More
Thermo Scientific™ μDrop™ and μDrop Duo Plates
Thermo Scientific™ μDrop and μDrop Duo plates are used for low volume (2-10 μl) photometric DNA, RNA and protein quantitation and purity analysis. The μDrop plates are compatible with Thermo Scientific Varioskan LUX and Multiskan SkyHigh microplate readers. The ready-made protocols in SkanIt software or Multiskan SkyHigh user interface make quantification fast and easy.
Brand:  Thermo Scientific™ N12391
2793.00 EUR valid until 2025-12-31
Use promo code "25043" to get your promotional price.
 View more versions of this product
Product Code. 11899143
  • 3145.00 € / Each
Explore more special offers
Description
Description
Thermo Scientific™ μDrop™ and μDrop™ Duo Plates are ideal tools for quick and easy photometric low-volume (2–10 μl) concentration measurements of DNA, RNA, protein samples, or spectral scanning. The low volume area is designed to be used with 8-channel pipette for fast and accurate multiple sample pipetting. The plates also contain a separate area for cuvette measurement. μDrop Plates are compatible with Thermo Scientific Varioskan™ LUX, Multiskan™ Sky and Multiskan SkyHigh microplate readers. The intuitive Thermo Scientific SkanIt™ software and Multiskan SkyHigh user interface offer ready-made measurement protocols for quick sample measurement and analysis.
  • dsDNA detection range from a few micrograms to a few milligrams per milliliter
  • Sixteen sample positions for quick and easy measurement of sample volumes down to 2μL
  • Compatible with an eight-channel pipette for easy sample addition
  • Quick and easy to wipe off the samples in serial measurements
  • Ready-made Thermo Scientific SkanIt Software sessions for nucleic acid analysis
  • Measurement slot for a rectangular cuvette
  • Compatible with Multiskan Sky, Multiskan GO, Varioskan LUX and Varioskan Flash
TRUSTED_SUSTAINABILITY
Specifications
Specifications
μDrop Plate
μDrop Duo Plate
2 to 10μL (low-volume function)
16
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ μDrop™ and μDrop Duo Plates >