Learn More
LABPLAS Dry Polyurethane Sani-Stick
Biocide free dry sponge with handle for sampling surfaces.
Brand:  LABPLAS KSS-67910
 View more versions of this product
Product Code. 17900205
  • 249.00 € / Pack of 100
Explore more special offers
Description
Description
  • Sani-Stick
  • Dry polyurethane sponge with handle
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Option to include gloves with kit
Specifications
Specifications
Hygiene Sponge Sampling Kit
Sterile
None
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
LABPLAS Dry Polyurethane Sani-Stick > None