Learn More
Welch Ilmvac™ Diaphragm Vacuum Pump Service and Maintenance Kit
Service and Maintenance Kits comprised of replacement seals and wearable parts for various models of diaphragm pumps.
Brand:  Welch Ilmvac™ 402042
 View more versions of this product
Product Code. 10090972
  • 518.00 € / Each
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available for a range of diaphragm vacuum pump models
  • Provided individually
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pump Models MP / MPC 601 T, 901 Z, 1201 E, MP 301V
Diaphragm, Valves And O-Rings.
1 Pack
Service Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Welch Ilmvac™ Diaphragm Vacuum Pump Service and Maintenance Kit > For Use With: Pump Models MP / MPC 601 T, 901 Z, 1201 E, MP 301V