Learn More
Thermo Scientific™ Imperial™ Protein Stain
Detect proteins in the low-nanogram range with this ready-to-use, in-gel, colorimetric stain formulated to deliver fast and sensitive SDS-PAGE staining.
Brand: Thermo Scientific™ 24615
144.40 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Thermo Scientific™ Imperial Protein Stain is a unique, ready-to-use colorimetric stain formulated with coomassie dye R-250 that delivers fast and sensitive total protein staining performance for low nanogram-level detection of proteins in polyacrylamide electrophoresis gels.
Imperial Protein Stain is a coomassie dye reagent for detection of protein bands in sodium dodecyl sulfate polyacrylamide gel electrophoresis (SDS-PAGE) and 2D gels. The stain is a unique formulation of coomassie brilliant blue R-250 that delivers substantial improvements in protein-staining performance compared to homemade or other commercial stains. Staining results in intensely colored protein bands that are easy to photograph and document with gel imagers. This reagent is one of the most sensitive colorimetric stains available, easily detecting 3 to 6 nanograms of protein per band. The Imperial Protein Stain protocol uses simple water-wash steps rather than methanol/acetic acid fixation and destaining, which saves valuable preparation time and minimizes reagent costs.Highlights:
Sensitive – detects less than 3ng protein per band with the enhanced protocol (3 hours)
Fast – ready-to-use reagent detects less than 6ng protein per band in just 20 minutes
Robust – highly consistent, reproducible protein staining technique
High contrast – intense purple bands are easier to photograph or scan than typical coomassie blue stains
Versatile – compatible with downstream mass spectrometry analysis and protein sequencing
Convenient – water washes only; no acid-fixative or methanol destaining solutions required
Stable – 1-year room-temperature stability ensures consistent performance and saves refrigerator space
Flexible – adjust staining and washing times to meet time- and sensitivity-requirements

Specifications
Upon receipt store product at room temperature. | |
Colorimetric | |
Protein Stain | |
1 L |
In-Blot Detection, In-Gel Detection | |
Coomassie | |
Imperial™ | |
Protein |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- IMPERIAL PROTEIN STAIN, 1 L
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
Supplemental information
- MIXTURE LIST-Contains : Acetic acid, Ethyl alcohol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZJ401489 | Certificate of Analysis | 16/12/2024 | 24615,24615_LITL18 PROMO |
ZI399875 | Certificate of Analysis | 02/11/2024 | 24615,24615_LITL18 PROMO |
ZI398306 | Certificate of Analysis | 15/10/2024 | 24615,24615_LITL18 PROMO |
ZG397509 | Certificate of Analysis | 22/08/2024 | 24615,24615_LITL18 PROMO |
ZG394855 | Certificate of Analysis | 07/08/2024 | 24615,24615_LITL18 PROMO |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.