missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Standard Taper Socket Side Arm, Unground Neck, without Stopper
Aspirator bottle with ground joint glass side-arm.
75.10 € - 478.00 €
Specifications
For Use With (Application) | Storage and dispensing of liquids and reagents |
---|---|
Material | Borosilicate Glass (3.3) |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10179350
|
DWK Life Sciences
247016302 |
2000 mL | N/A | N/A | N/A | |||||
15231999
|
DWK Life Sciences
247015409 |
1000 mL | N/A | N/A | N/A | |||||
15221999
|
DWK Life Sciences
247014404 |
500 mL | N/A | N/A | N/A | |||||
Description
Aspirator bottles, ground glass side-arm fro dispensing of liquids. Complying with ISO 4796, these aspirators have a ground socket side-arm and plain neck to accept rubber stoppers. Compatible stopcocks assemblies are available separately.
- Outlet facilitates attachment of ground jointed stopcock
- With plain unground neck for rubber stopper and bottom ground joint outlet
- Made of virtually inert, chemically resistant DURAN™ glass
Specifications
Storage and dispensing of liquids and reagents | |
Borosilicate Glass (3.3) | |
Autoclavable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Standard Taper Socket Side Arm, Unground Neck, without Stopper