Learn More
Dysprosium(III) perchlorate hexahydrate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals
51.70 € - 148.00 €
Chemical Identifiers
CAS | 14692-17-2 |
---|---|
Molecular Formula | Cl3DyO12 |
Molecular Weight (g/mol) | 460.838 |
MDL Number | MFCD00150227 |
InChI Key | QNDHOIIJUAAORW-UHFFFAOYSA-K |
Synonym | dysprosium 3+ perchlorate, dysprosium iii perchlorate, dysprosium perchlorate, acmc-1c5o7, dysprosium iii perchlorate solution, perchloric acid dysprosium iii salt, dysprosium 3+ triperchlorate, perchloric acid,dysprosium 3+ salt 8ci,9ci, perchloric acid,dysprosium 3+ salt, hexahydrate 8ci,9ci |
PubChem CID | 13767046 |
IUPAC Name | dysprosium(3+);triperchlorate |
SMILES | [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Dy+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11440301
|
Thermo Scientific Alfa Aesar
040574.NA |
2 g |
51.70 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450301
|
Thermo Scientific Alfa Aesar
040574.NC |
10 g | |||||||
Description
A solution of 2,6-diformyl-4-methylphenol (0.15mmol) and benzoylhydrazide (0.30 mmol) in MeOH (10 ml) was allowed to react with the dysprosium(III) perchlorate hexahydrate (0.3 mmol) in 10 ml MeOH In observation of slow magnetic relaxation in triple-stranded lanthanide helicates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14692-17-2 | |
460.838 | |
QNDHOIIJUAAORW-UHFFFAOYSA-K | |
13767046 | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Dy+3] |
Cl3DyO12 | |
MFCD00150227 | |
dysprosium 3+ perchlorate, dysprosium iii perchlorate, dysprosium perchlorate, acmc-1c5o7, dysprosium iii perchlorate solution, perchloric acid dysprosium iii salt, dysprosium 3+ triperchlorate, perchloric acid,dysprosium 3+ salt 8ci,9ci, perchloric acid,dysprosium 3+ salt, hexahydrate 8ci,9ci | |
dysprosium(3+);triperchlorate |
Specifications
14692-17-2 | |
Cl3DyO12 | |
UN3211 | |
Insoluble in water. | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Dy+3] | |
460.838 | |
568.94 (460.85 Anhydrous) | |
50% w/w aq. soln. | |
Dysprosium(III) perchlorate hexahydrate |
Liquid | |
MFCD00150227 | |
dysprosium 3+ perchlorate, dysprosium iii perchlorate, dysprosium perchlorate, acmc-1c5o7, dysprosium iii perchlorate solution, perchloric acid dysprosium iii salt, dysprosium 3+ triperchlorate, perchloric acid,dysprosium 3+ salt 8ci,9ci, perchloric acid,dysprosium 3+ salt, hexahydrate 8ci,9ci | |
QNDHOIIJUAAORW-UHFFFAOYSA-K | |
dysprosium(3+);triperchlorate | |
13767046 | |
Reagent | |
Odorless |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H271-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
missing translation for 'einecsNumber' : 237-838-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only