Learn More
Bellingham + Stanley™ Eclipse Hand Held Refractometer
Manufactured to the highest specification using only the best materials and optical components. Bellingham + Stanley™ Eclipse Hand Held Refractometer is a low-cost, portable instrument used for on-the-spot determination of Refractive Index of liquids or the concentration of solutions.
Brand:  Bellingham + Stanley™ 45-01
 View more versions of this product
Product Code. 12303519
  • 303.00 € / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Eclipse professional-grade range of scales are available for general use in a variety of applications, with special models available, such as Low Volume Brix models to facilitate measurement of samples of less than 1μL that are used by botanists and zoologists for measuring nectar. All instruments are supplied with a soft carrying case and instructions for use.
These reliable and robust refractometers are sealed against the ingress of water with IP65 construction, so they can be used in the harshest of environments. The large body housing and rubber handgrip make the instrument incredible easy to handle. Special features make measurements easy and reliable:
  • Simple zero adjust with lock
  • Shaded view with bright display for maximum distinction
  • High precision, clear scale
  • Easy-clean prism
  • Unique sample ‘dribble feed’ feature
  • Low weight
  • Anti-roll supports
  • Push-on prism flap
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Eclipse Professional Grade Optical Hand Held Refractometer
0.1
‘On the spot’ determination of Refractive Index (RI) of liquids or the concentration of solutions
IP65
0 to 15 Brix
Brix
Soft carrying case
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bellingham + Stanley™ Eclipse Hand Held Refractometer > Sugar % Concentration; Brix Range: 0-15%; Increments: 0.1