missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™ 96-Well Assay Plates for Cell-Based Assays

Offer formats that allow a wide range of functional cell-based and whole-organism assays to be performed

Brand:  Merck MAMIC5S10

 View more versions of this product

Product Code. 10486892

  • 1800.00 € / Pack of 10
Estimated Shipment: 31-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Ideal for cell viability and proliferation, migration, invasion, and chemotaxis assays. Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
8 x 12
Clear
96
Tissue Culture
Migration, invasion and chemotaxis
Polycarbonate
5 μm
Styrene
Sterile
0.28cm2
10/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Merck MultiScreen™ 96-Well Assay Plates for Cell-Based Assays > Multscreen MIC; 10/pk; 5.0μm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.