Learn More
Eppendorf epT.I.P.S.™ Motion as Reload System
BIOPHARMAGREENER_CHOICE
Eppendorf epT.I.P.S. Motion work with the epMotion platform, with a TipHolder accessory the reload trays offer an environmentally friendly option.
Brand:  Eppendorf 0030014430
486.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 30019045
  • 492.00 € / Pack of 2304
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • epT.I.P.S. Motion can be used with epMotion™ 96 or integrated into your workstation as a reload system when combined with the tip holder
  • The high-quality reload tips are delivered in a PET thermoformed tray with sealed lid. Upgrading to the autoclavable, aluminum tip holder, which replaces the disposable box, provides waste reduction of up to 40%
  • Eppendorf consumables and thus also epT.I.P.S. Motion pipette tips as Reload System are shipped in packaging consisting of at least 62% recycled cardboard
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.5 to 50 μL
PCR Clean
Not Autoclavable
Hanging Tip Rack
Robotic Tips
Eppendorf epMotion
Polypropylene
Filtered
Clear
For Eppendorf epMotion series
Standard, Straight
Single-use
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf epT.I.P.S.™ Motion as Reload System > Volume (Metric): 0.5 - 50 μL; Filtered: Filtered; Sterility: PCR clean