missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 2-amino-6-Boc-4,7-dihydrothieno[2,3-c]pyridine-3(5H)-carboxylate, 97%, Thermo Scientific™ Product Code.: 15438865

Ethyl 2-amino-6-Boc-4,7-dihydrothieno[2,3-c]pyridine-3(5H)-carboxylate, 97%, Thermo Scientific™

Product Code. 15438865
5 g
Click to view available options
Quantity:
250 mg
5 g
This item is not returnable. View return policy

Product Code. 15438865

Brand: Thermo Scientific Alfa Aesar H34279.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193537-14-3
Molecular Formula C15H22N2O4S
Molecular Weight (g/mol) 326.411
MDL Number MFCD05664039
InChI Key XSXVOVXVHBSSSN-UHFFFAOYSA-N
Synonym ethyl 6-boc-2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3-carboxylate, 2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3,6-dicarboxylic acid 6-tert butyl ester 3-ethyl ester, 6-tert-butyl 3-ethyl 2-amino-4,5-dihydrothieno 2,3-c pyridine-3,6 7h-dicarboxylate, 6-tert-butyl 3-ethyl 2-amino-4h,5h,6h,7h-thieno 2,3-c pyridine-3,6-dicarboxylate, 6-tert-butyl 3-ethyl 2-amino-4,7-dihydrothieno 2,3-c pyridine-3,6 5h-dicarboxylate, 2-amino-6-boc-4,5,6,7-tetrahydro-thieno 2,3-c pyridine-3-carboxylic acid ethyl ester, 2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3,6-dicarboxylic acid 6-tert-butyl ester 3-ethyl ester, 6-tert-butyl 3-ethyl 2-amino-4h,5h,7h-thieno 2,3-c pyridine-3,6-dicarboxylate, ethyl 2-amino-4,5,6,7-tetrahydrothieno 2,3-c pyridine-3-carboxylate, n-boc protectedShow MoreShow Less
PubChem CID 2794740
IUPAC Name 6-O-tert-butyl 3-O-ethyl 2-amino-5,7-dihydro-4H-thieno[2,3-c]pyridine-3,6-dicarboxylate
SMILES CCOC(=O)C1=C(SC2=C1CCN(C2)C(=O)OC(C)(C)C)N

Specifications

Melting Point 151°C to 153°C
Quantity 5 g
Formula Weight 326.42
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-6-Boc-4,7-dihydrothieno[2,3-c]pyridine-3(5H)-carboxylate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10171768 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 2-amino-6-Boc-4,7-dihydrothieno[2,3-c]pyridine-3(5H)-carboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.