missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™

61.20 € - 328.00 €

Chemical Identifiers

CAS 7238-62-2
Molecular Formula C7H8ClNO2S
Molecular Weight (g/mol) 205.656
InChI Key VUARUZUFHDNJSY-UHFFFAOYSA-N
Synonym ethyl 2-chloro-4-methylthiazole-5-carboxylate, ethyl 2-chloro-4-methyl-5-thiazolecarboxylate, 5-thiazolecarboxylic acid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-5-thiazolecarboxylic acid ethyl ester, 5-thiazolecarboxylicacid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-thiazole-5-carboxylic acid ethyl ester
PubChem CID 594903
IUPAC Name ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)Cl)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10127302
View Documents Promotion Details
Thermo Scientific Maybridge
MO07617CB
250 mg
61.20 €
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10516693
View Documents Promotion Details
Thermo Scientific Maybridge
MO07617DA
1 g
82.10 €
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10492674
View Documents Promotion Details
Thermo Scientific Maybridge
MO07617DE
5 g
328.00 €
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7238-62-2
205.656
ethyl 2-chloro-4-methylthiazole-5-carboxylate, ethyl 2-chloro-4-methyl-5-thiazolecarboxylate, 5-thiazolecarboxylic acid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-5-thiazolecarboxylic acid ethyl ester, 5-thiazolecarboxylicacid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-thiazole-5-carboxylic acid ethyl ester
ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate
C7H8ClNO2S
VUARUZUFHDNJSY-UHFFFAOYSA-N
594903
CCOC(=O)C1=C(N=C(S1)Cl)C

Specifications

7238-62-2
C7H8ClNO2S
VUARUZUFHDNJSY-UHFFFAOYSA-N
ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate
594903
97%
97%
ethyl 2-chloro-4-methylthiazole-5-carboxylate, ethyl 2-chloro-4-methyl-5-thiazolecarboxylate, 5-thiazolecarboxylic acid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-5-thiazolecarboxylic acid ethyl ester, 5-thiazolecarboxylicacid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-thiazole-5-carboxylic acid ethyl ester
CCOC(=O)C1=C(N=C(S1)Cl)C
205.656
205.66
Ethyl 2-chloro-4-methyl-1, 3-thiazole-5-carboxylate
Videos
SDS
Documents

Documents

Product Certifications